Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbetapentane citrate, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
A selective ligand for the σ1 -site.
Specifications
Quantity | 100 mg |
Formula Weight | 525.59 |
Percent Purity | >99% |
Chemical Name or Material | Carbetapentane citrate |
Safety and Handling
Recommended Storage : Store at Room Temperature
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title