Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Carbethoxyethylidene)triphenylphosphorane, ca. 94%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
5717-37-3
C23H23O2P
362.4
MFCD00009160
KZENFXVDPUMQOE-UHFFFAOYSA-N
ethyl 2-triphenylphosphoranylidene propanoate,carbethoxyethylidene triphenylphosphorane,ethyl 2-triphenylphosphoranylidene propionate,1-ethoxycarbonylethylidene triphenylphosphorane,propanoic acid, 2-triphenylphosphoranylidene-, ethyl ester,1-ethoxycarbonyl ethylidenetriphenylphosphorane,ethyl 2-triphenyl-lambda5-phosphanylidene propanoate,ethyl 2-triphenyl-$l^ 5-phosphanylidene propanoate,1-ethoxycarbonylethylethylidene triphenylphosporane,carbethoxyethylidine triphenylphosphorane
79792
ethyl 2-(triphenyl-$l^{5}-phosphanylidene)propanoate
CCOC(=O)C(=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C
5717-37-3
C23H23O2P
362.4
MFCD00009160
KZENFXVDPUMQOE-UHFFFAOYSA-N
ethyl 2-triphenylphosphoranylidene propanoate,carbethoxyethylidene triphenylphosphorane,ethyl 2-triphenylphosphoranylidene propionate,1-ethoxycarbonylethylidene triphenylphosphorane,propanoic acid, 2-triphenylphosphoranylidene-, ethyl ester,1-ethoxycarbonyl ethylidenetriphenylphosphorane,ethyl 2-triphenyl-lambda5-phosphanylidene propanoate,ethyl 2-triphenyl-$l^ 5-phosphanylidene propanoate,1-ethoxycarbonylethylethylidene triphenylphosporane,carbethoxyethylidine triphenylphosphorane
79792
ethyl 2-(triphenyl-$l^{5}-phosphanylidene)propanoate
CCOC(=O)C(=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C
Packaging Glass bottle
Melting Point 157.0°C to 159.0°C
Quantity 25g
Color Yellow
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range ca. 94%
Linear Formula CH3C[=P(C6H5)3]CO2C2H5
Formula Weight 362.4
Physical Form Crystalline Powder
Percent Purity ca. 94%
Chemical Name or Material (Carbethoxyethylidene)triphenylphosphorane, Balance TPPO
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger