Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbic anhydride, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
Packaging:
Glass bottle
129-64-6
C9H8O3
164.16
MFCD00151106
KNDQHSIWLOJIGP-USEPMLCBSA-N
cis-5-norbornene-endo-2,3-dicarboxylic anhydride
126963424
C1C2C=CC1C3C2C(=O)OC3=O
129-64-6
C9H8O3
164.16
MFCD00151106
KNDQHSIWLOJIGP-USEPMLCBSA-N
cis-5-norbornene-endo-2,3-dicarboxylic anhydride
126963424
C1C2C=CC1C3C2C(=O)OC3=O
Specific Gravity 1.08
Melting Point 162.0°C to 165.0°C
Quantity 100g
Packaging Glass bottle
Density 1.0800g/mL
Infrared Spectrum Authentic
Refractive Index 1.4365
Color White
Assay Percent Range Balance: himic acid
Merck Index 15, 1797
Solubility Information Solubility in water: <0,01g/100g. Other solubilities: soluble in acetone,benzene,ethyl ether,ethanol,decomposes with alkalies
Formula Weight 164.16
Physical Form Crystalline Powder
Percent Purity 99+%
Chemical Name or Material Carbic anhydride
Show More Show Less

GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of s

GHS Signal Word: Danger

EINECSNumber : 204-957-7