Learn More
Medchemexpress LLC Carbidopa | 28860-95-9 | 99.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB03115MG
Encompass_Preferred
Carbidopa is a peripheral decarboxylase inhibitor used in the research of Parkinson's disease. It acts as a selective aryl hydrocarbon receptor (AhR) modulator and has been shown to inhibit pancreatic cancer cell and tumor growth.
  • It is a peripheral decarboxylase inhibitor.
  • It can be used for the research of Parkinson's disease.
  • It acts as a selective aryl hydrocarbon receptor modulator.
  • It inhibits pancreatic cancer cell and tumor growth.
Catalog No. 50-004-06358