Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbidopa monohydrate, 99%, Acros Organics™

Click to view available options
Quantity:
1g
38821-49-7
C10H14N2O4·H2O
244.25
QTAOMKOIBXZKND-PPHPATTJSA-N
3-3,4-dihydroxyphenyl-2-hydrazinyl-2-methylpropanoic acid hydrate,+-2-3,4-dihydroxybenzyl-2-hydrazinopropionic acid monohydrate,3-3,4-bis oxidanyl phenyl-2-diazanyl-2-methyl-propanoic acid hydrate
38101
CHEBI:3395
(2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid;hydrate
CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)NN.O
38821-49-7
C10H14N2O4·H2O
244.25
QTAOMKOIBXZKND-PPHPATTJSA-N
3-3,4-dihydroxyphenyl-2-hydrazinyl-2-methylpropanoic acid hydrate,+-2-3,4-dihydroxybenzyl-2-hydrazinopropionic acid monohydrate,3-3,4-bis oxidanyl phenyl-2-diazanyl-2-methyl-propanoic acid hydrate
38101
CHEBI:3395
(2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid;hydrate
CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)NN.O
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Loss on Drying 6.9% to 7.9% (1 g, 105°C)
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 244.25
Percent Purity 99%
Chemical Name or Material Carbidopa monohydrate

GHS H Statement
Causes serious eye irritation.

GHS P Statement Wear eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling. If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning