Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbidopa, USP, 98-102%, Spectrum™ Chemical
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Description
Spectrum™ Chemical Carbidopa, USP is used in combination with levodopa to treat symptoms of Parkinson′s disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Quantity | 1 g |
Percent Purity | 98 to 102% |
Grade | USP |
Ignition Residue | 0.1% |
Loss on Drying | 6.9-7.9% |
Packaging | Amber Glass Bottle |
Specific Rotation | -21.0° to -23.5° |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction