Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbidopa, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
38821-49-7
C10H16N2O5
244.25
QTAOMKOIBXZKND-PPHPATTJSA-N
(2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid hydrate
O.C[C@@](CC1=CC=C(O)C(O)=C1)(NN)C(O)=O
38821-49-7
C10H16N2O5
244.25
QTAOMKOIBXZKND-PPHPATTJSA-N
(2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid hydrate
O.C[C@@](CC1=CC=C(O)C(O)=C1)(NN)C(O)=O

C1401, 38821-49-7, C10H14N2O4•H2O

Spectrum™ Carbidopa, USP is used in combination with levodopa to treat symptoms of Parkinson′s disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.1%
Percent Purity 98-102%
Loss on Drying 6.9-7.9%
Packaging Amber Glass Bottle
Specific Rotation -21.0° to -23.5°
Quantity 1 kg