Learn More
Carbidopa, USP, 98-102%, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Poly Bottle
C1401, 38821-49-7, C10H14N2O4•H2O
Spectrum™ Carbidopa, USP is used in combination with levodopa to treat symptoms of Parkinson′s disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.001
Loss on Drying 6.9-7.9%
Packaging Amber Glass Bottle
Specific Rotation -21.0° to -23.5°