Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbohydrate Sulfotransferase 2/CHST2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179191
Description
Carbohydrate Sulfotransferase 2/CHST2 Polyclonal specifically detects Carbohydrate Sulfotransferase 2/CHST2 in Mouse samples. It is validated for Western Blot.Specifications
Carbohydrate Sulfotransferase 2/CHST2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
C6ST, carbohydrate (chondroitin 6/keratan) sulfotransferase 2, carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2, carbohydrate sulfotransferase 2, EC 2.8.2.-, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2, glcNAc6ST-1, GN6ST, Gn6ST-1, GST2, GST-2, N-acetylglucosamine 6-O-sulfotransferase 1 | |
Rabbit | |
Affinity purified | |
RUO | |
9435 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_061233 | |
CHST2 | |
The specific Immunogen is proprietary information. Peptide sequence VFQLYSPAGSGGRNLTTLGIFGAATNKVVCSSPLCPAYRKEVVGLVDDRV. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction