Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbohydrate Sulfotransferase 6/CHST6 Mouse anti-Human, Mouse, Canine, Monkey, Alexa Fluor™ 647, Clone: OTI1D1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271195AF647

Catalog No. N271195A647


Add to cart

Description

Description

Carbohydrate Sulfotransferase 6/CHST6 Monoclonal antibody specifically detects Carbohydrate Sulfotransferase 6/CHST6 in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot.
Specifications

Specifications

Carbohydrate Sulfotransferase 6/CHST6
OTI1D1
Western Blot
Affinity Purified, Conjugated
CHST6
Recombinant protein fragment corresponding to amino acids 202-395 of human Carbohydrate Sulfotransferase 6/CHST6 (NP_067628) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey, Mouse
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6, carbohydrate sulfotransferase 6, C-GlcNAc6ST, corneal N-acetylglucosamine 6-sulfotransferase, Corneal N-acetylglucosamine-6-O-sulfotransferase, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.17, EC 2.8.2.21, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta, glcNAc6ST-5, Gn6st-5, GST4-beta, hCGn6ST, MCDC1, N-acetylglucosamine 6-O-sulfotransferase 5
Mouse
IgG1
0.1 mL
Primary
4166.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit