Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbohydrate Sulfotransferase 6/CHST6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Carbohydrate Sulfotransferase 6/CHST6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157927
|
Novus Biologicals
NBP157927 |
100 μL |
Each for $436.00
|
|
NBP15792720
|
Novus Biologicals
NBP15792720UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Carbohydrate Sulfotransferase 6/CHST6 Polyclonal specifically detects Carbohydrate Sulfotransferase 6/CHST6 in Human samples. It is validated for Western Blot.Specifications
Carbohydrate Sulfotransferase 6/CHST6 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6, carbohydrate sulfotransferase 6, C-GlcNAc6ST, corneal N-acetylglucosamine 6-sulfotransferase, Corneal N-acetylglucosamine-6-O-sulfotransferase, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.17, EC 2.8.2.21, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta, glcNAc6ST-5, Gn6st-5, GST4-beta, hCGn6ST, MCDC1, N-acetylglucosamine 6-O-sulfotransferase 5 | |
CHST6 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q9GZX3 | |
4166 | |
Synthetic peptides corresponding to CHST6(carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6) The peptide sequence was selected from the middle region of CHST6. Peptide sequence YAFTGLSLTPQLEAWIHNITHGSGPGARREAFKTSSRNALNVSQAWRHAL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title