Alfa Aesar™ Carbohydrazide, 97%

Manufacturer: Alfa Aesar™ A1114522

Quantity 100g

 View more versions of this product

Catalog No. AAA1114522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 497-18-7
    Molecular Formula CH6N4O
    Linear Formula (H2NNH)2C=O
    Formula Weight 90.09
    MDL Number MFCD00007591
    UN Number UN3077
    Merck Index 14,1804
    Chemical Name or Material Carbohydrazide

    Specifications

    Formula Weight 90.09
    Health Hazard 3 GHS P Statement
    P280h-P273-P305+P351+P338-P308+P313-P330-P501a
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or concerned: Get medical advice/attention.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 497-18-7
    UN Number UN3077
    RTECS Number FF2625000
    Quantity 100g
    Molecular Formula CH6N4O
    Linear Formula (H2NNH)2C=O
    MDL Number MFCD00007591
    Merck Index 14,1804
    Health Hazard 1 Warning
    Recommended Storage 2°C to 8°C
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Melting Point 153° to 158°C
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Carbohydrazide
    Assay 97%

    Description

    Applications
    Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.

    Solubility
    Soluble in water.

    Notes
    Store in cool place. Incompatible with strong oxidizing agents, strong acids, copper, zinc, nickel, lead and brass.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P280h-P273-P305+P351+P338-P308+P313-P330-P501a
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or concerned: Get medical advice/attention.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit