Learn More
Carbomer 934, Resin, Powder, NF, Spectrum™
Click to view available options
Packaging:
Fiber Drum
CA251, 9003-01-4
Spectrum™ Carbomer 934, Resin, Powder, NF polymers can provide bioadhesion, taste-masking and good binding characteristics that provide highly effective controlled-release properties. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 0.02
Packaging Fiber Drum