Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbomer 940, Thickening Agent, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
9003-01-4
9003-01-4
CAS Max % 1
Heavy Metals 1e-05
Grade Reagent
Packaging Poly Bottle
Water 0.02
Quantity 100 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.