Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbomer 941, NF, Spectrum™

Click to view available options
Quantity:
100 g
2.5 kg
45 kg
50 lbs.
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1/4/9003
1/4/9003

C1478, 9003-01-4

Spectrum™ Carbomer 941, NF is an expanded molecule that will increase continually when more alkyne bonds are presented. It is because of this that carbomers are also called carbon-molecules. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Loss on Drying 2%
Packaging Fiber Drum
Quantity 50 lbs.