Learn More
Carbomer 941, Thickening Agent, Spectrum™
Click to view available options
Packaging:
Poly Bottle
Poly Pail
C1186, 9003-01-4
Spectrum™ Carbomer 941, Thickening Agent gives permanent emulsions and suspensions at low viscosity, even with ionic systems. The gels produced with this polymer have excellent clarity. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Poly Bottle