Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon, Activated, Powder, J.T.Baker™

Click to view available options
Quantity:
500g
3kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7440-44-0
C
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
DARCO™G-60
5462310
CHEBI:27594
carbon
[C]
Quantity 500g
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 12.01
Physical Form Powder
Chemical Name or Material Carbon, Activated