Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbon black, acetylene, >99.9%, Alfa Aesar™
Click to view available options
Quantity:
250g
1kg
3kg
Specifications
Color | Black |
Odor | Odorless |
Density | Bulk density 80 to 120g/L |
Quantity | 250g |
UN Number | UN1325 |
Physical Form | S.A. 75m |
Percent Purity | ≥99.9% |
Chemical Name or Material | Carbon black, acetylene, 50% compressed |
Safety and Handling
GHS H Statement
H228-H351;
Flammable solid.
Suspected of causing cancer.
P201-P202-P281-P308+P313-P501c
H351
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 215-609-9
RTECSNumber : FF5800000
TSCA : Yes
Recommended Storage : Ambient temperatures