Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon, Darco G-60, Activated, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
500g
Packaging:
Glass Bottle
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite,activated charcoal,norit,mineral,carbon-12,carbono,graphene,acticarbone,anthrasorb,carbosieve
5462310
CHEBI:27594
carbon
[C]
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite,activated charcoal,norit,mineral,carbon-12,carbono,graphene,acticarbone,anthrasorb,carbosieve
5462310
CHEBI:27594
carbon
[C]

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Packaging Glass Bottle
Density 1.8g/mL
Color Black
Melting Point 3652°C
Vapor Pressure 1mmHg at 3586°C
Quantity 500g
Identification Pass Test
Formula Weight 12.01
Physical Form Solid
Chemical Name or Material Carbon, Darco™ G-60, Activated

CAUTION!

Emergency Overview
May cause skin and eye irritation. Avoid contact with skin and eyes. Use personal protective equipment. Look out! DATATYPE 710. Avoid breathing dust/fume/gas/mist/vapours/spray. Rinse with water. . Obtain medical attention. Remove from exposure, lie down. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0