Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Disulfide, ACS, 99.9%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
75-15-0
75-15-0
CAS Max % 1
Percent Purity 0.999
Grade ACS
Hydrogen Sulfide To Pass Test
Packaging Amber Glass Bottle
Residue after Evaporation 2e-05
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.