Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Disulfide, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Aluminum Bottle

Chemical Identifiers

75-15-0
CS2
76.131
QGJOPFRUJISHPQ-UHFFFAOYSA-N
6348
CHEBI:23012

High quality chemicals for laboratory and specialized industrial use

CAS 75-15-0
Molecular Formula CS2
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILES C(=S)=S
IUPAC Name methanedithione
Molecular Weight (g/mol) 76.131
PubChem CID 6348
ChEBI CHEBI:23012
Formula Weight 76.13
Packaging Aluminum Bottle
Specific Gravity 1L = 1.26kg
Flash Point −30.00°C
Quantity 2.5L
Chemical Name or Material Carbon Disulfide
Grade Baker Analyzed™ ACS Reagent
Show More Show Less