Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Disulfide, Technical, Spectrum™

Click to view available options
Quantity:
500 mL
2.5 L
18 L
20 L
Packaging:
Amber Glass Bottle
Poly-Coated Glass Bottle
Steel Can
75-15-0
75-15-0
CAS Max % 1
Grade Technical
Packaging Amber Glass Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.