Alfa Aesar™ Carbon felt, 2.54cm (1.0 in.) thick, 99.0%

Manufacturer: Alfa Aesar™ 43201RR

Quantity 10 x 10cm

 View more versions of this product

Catalog No. AA43201RR

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00133992
    Chemical Name or Material Carbon felt, 2.54cm (1.0 in.) thick

    Specifications

    Quantity 10 x 10cm
    Weight ≈19.1g/10x10cm
    MDL Number MFCD00133992
    Chemical Name or Material Carbon felt, 2.54cm (1.0 in.) thick
    Assay 99.0%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit