Learn More
Justrite™ Carbon Filter Replacement Indicator
For Use With: VaporTrap Solvent Waste Systems
Supplier: Justrite™ 12952
Description
- Qualitative colormetric indicator
- Ensures proper filter functionality with end of service life indicator
- Provide real-time indication of breakthrough of HPLC solvents vapors in filter
- Indicator changes color to red when fil
Specifications
L x Dia.: 3.1 x 0.4 in. | |
Carbon | |
Indicator | |
VaporTrap Solvent Waste Systems |