Alfa Aesar™ Carbon powder, activated, Norit™ D10

Manufacturer: Alfa Aesar™ H36691-0P

Quantity 2000g

 View more versions of this product

Catalog No. AAH366910P

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-44-0
    MDL Number MFCD00133992
    UN Number UN1362
    Chemical Name or Material Carbon powder, Activated, Norit™ D10

    Specifications

    MDL Number MFCD00133992
    UN Number UN1362
    Health Hazard 2 GHS H Statement
    H228-H252
    Flammable solid.
    Self-heating in large quantities
    may catch fire.
    Health Hazard 3 GHS P Statement
    P210-P280-P235+P410-P240-P370+P378a-P420
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep cool.
    Protect from su
    TSCA TSCA
    Physical Form Powder
    Color Black
    Quantity 2000g
    CAS 7440-44-0
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packaging Group: III; Proper Shipping Name: CARBON, ACTIVATED
    RTECS Number FF5250100
    Chemical Name or Material Carbon powder, Activated, Norit™ D10

    GHS H Statement
    H228-H252
    Flammable solid.
    Self-heating in large quantities
    may catch fire.

    GHS P Statement
    P210-P280-P235+P410-P240-P370+P378a-P420
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep cool.
    Protect from su

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit