Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbon Standard (Organic), 1000ppm, Ricca Chemical

Click to view available options
Quantity:
500mL
Packaging:
Glass amber
CAS 877-24-7, 7664-38-2, 877-24-7, 7732-18-5
CAS Min % 97.0, 0.46, 0.21
CAS Max % 100.0, 0.48, 0.22
Molecular Formula C8H5KO4
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
IUPAC Name potassium;2-carboxybenzoate
Molecular Weight (g/mol) 204.222
PubChem CID 23676735
Physical Form Liquid
Packaging Glass amber
Equivalence 1 mL = 1 mg C
Color Colorless
Quantity 500mL
Concentration or Composition Notes 1000ppm
Chemical Name or Material Organic Carbon Standard
Show More Show Less