Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.0
$89.00 - $121.65
Specifications
Concentration | 0.5 M buffer soln. |
---|---|
pH Range | 9.0 |
Type | Carbonate buffer soln. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ63899AK
|
Thermo Scientific Chemicals
J63899AK |
250 mL |
Each for $89.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ63899AP
|
Thermo Scientific Chemicals
J63899AP |
500 mL |
Each
for $121.65
Save 11% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
0.5 M buffer soln. | |
9.0 | |
Carbonate buffer soln. |
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title