Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Carbonic Anhydrase I/CA1 Human Recombinant Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Supplier: Novus Biologicals™ NBP2522410.1MG
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-261 of Human Carbonic Anhydrase I/CA1 The Recombinant Human Carbonic Anhydrase I/CA1 Protein is derived from E. coli. The Recombinant Human Carbonic Anhydrase I/CA1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
1mg/mL | |
20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol | |
TMW: 31kDa | |
0.1mg | |
CA-I, Car1, Carbonate dehydratase I, carbonic anhydrase 1, Carbonic anhydrase B, carbonic anhydrase ICAB, carbonic dehydratase, EC 4.2.1.1 | |
Specific activity is > 300 pmol/min/ug, and is defined as the amount of enzyme that hydrolyze 1.0 pmole of 4-nitrophenyl acetate to 4-nitrophenol per minute at pH 8.0 at 37C. | |
Yes |
SDS-Page | |
759 | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
Carbonic Anhydrase I/CA1 | |
Unlabeled |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only and is not approved for use in humans or in clinical diagnosis