Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase VA/CA5A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen Carbonic Anhydrase VA/CA5A
Immunogen Synthetic peptides corresponding to CA5A (carbonic anhydrase VA, mitochondrial) The peptide sequence was selected from the C terminal of CA5A. Peptide sequence PSQLSAFRTLLFSALGEEEKMMVNNYRPLQPLMNRKVWASFQATNEGTRS.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16888920 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16888920UL
20ul Each for $149.00
Add to cart
 
NBP168889 View Documents Novus BiologicalsSupplier Diversity Partner
NBP168889
100 ul Each for $329.00
Add to cart
 
Description

Description

Carbonic Anhydrase VA/CA5A Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Carbonic Anhydrase VA/CA5A
Rabbit
Polyclonal
Unconjugated
763
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to CA5A (carbonic anhydrase VA, mitochondrial) The peptide sequence was selected from the C terminal of CA5A. Peptide sequence PSQLSAFRTLLFSALGEEEKMMVNNYRPLQPLMNRKVWASFQATNEGTRS.
Primary
IgG
CA5carbonic anhydrase 5A, mitochondrial, Carbonate dehydratase VA, carbonic anhydrase V, mitochondrial, Carbonic anhydrase VA, carbonic anhydrase VA, mitochondrial, carbonic dehydratase, CAV, CAVA, CA-VA, EC 4.2.1.1
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit