Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VB/CA5B Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP299971AF647
Description
Carbonic Anhydrase VB/CA5B Polyclonal antibody specifically detects Carbonic Anhydrase VB/CA5B in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
Carbonic Anhydrase VB/CA5B | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Carbonate dehydratase VB, carbonic anhydrase 5B, mitochondrial, Carbonic anhydrase VB, carbonic anhydrase VB, mitochondrial, carbonic dehydratase, CA-VB, EC 4.2.1.1, MGC39962 | |
Produced in rabbits immunized with purified, recombinant Carbonic Anhydrase VB/CA5B (Q9Y2D0; Cys34-Pro317). | |
Affinity purified | |
RUO | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
ELISA, Immunofluorescence | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
Lipid and Metabolism | |
Primary | |
11238 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title