Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XII/CA12 Antibody (08), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP326096MFV450
Description
Carbonic Anhydrase XII/CA12 Monoclonal antibody specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for ELISASpecifications
Carbonic Anhydrase XII/CA12 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
08 | |
ELISA | |
Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
0.1 mL | |
Lipid and Metabolism | |
771 | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction