Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase XII/CA12 Mouse anti-Human, Clone: OTI2C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270329

Catalog No. NBP270329


Add to cart

Description

Description

Carbonic Anhydrase XII/CA12 Monoclonal antibody specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Carbonic Anhydrase XII/CA12
OTI2C6
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3
Mouse
IgG1
Immunogen affinity purified
Lipid and Metabolism
Primary
771
Human
Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:500, Immunohistochemistry 1:150
Affinity Purified, Lyophilized
CA12
Full length human recombinant protein of human CA12(NP_001209) produced in HEK293T cell.
36.8 kDa
100 μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit