Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XII/CA12 Mouse anti-Human, DyLight 488, Clone: OTI2C6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270329G
Description
Carbonic Anhydrase XII/CA12 Monoclonal antibody specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Carbonic Anhydrase XII/CA12 | |
OTI2C6 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
CA12 | |
Full length recombinant protein of human Carbonic Anhydrase XII/CA12 (NP_001209) produced in HEK293T cells. | |
Immunogen affinity purified | |
Lipid and Metabolism | |
Primary | |
771.0 |
Immunohistochemistry, Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title