Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase XII Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537845
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Carbonic Anhydrase XII is a protein coding gene.
Specifications
Carbonic Anhydrase XII | |
Polyclonal | |
Unconjugated | |
CA12 | |
2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
771 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43570 | |
CA12 | |
Peptide with sequence C-SHLQHVKYKGQE, from the internal region of CA12 (aa188-199). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction