Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XIII Recombinant Rabbit Monoclonal Antibody (103), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA543470

This item is not returnable.
View return policy
Description
Specificity: Human Carbonic Anhydrase XIII/CA13
Carbonic Anhydrase XIII Recombinant Monoclonal antibody specifically detects Carbonic Anhydrase XIII in Human samples. It is validated for ELISASpecifications
Carbonic Anhydrase XIII | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q8N1Q1 | |
Ca13 | |
Recombinant Human Carbonic Anhydrase XIII/CA13 protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
103 | |
Unconjugated | |
Ca13 | |
2310075C21Rik; Ca13; Car13; Carbonate dehydratase XIII; carbonic anhydrase 13; Carbonic anhydrase XIII; CAXIII; CA-XIII; RGD1560453 | |
Rabbit | |
Protein A | |
RUO | |
377677 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title