Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase XIV/CA14 Rabbit anti-Human, Clone: 4, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | Carbonic Anhydrase XIV/CA14 |
---|---|
Clone | 4 |
Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Carbonic Anhydrase XIV/CA14 Monoclonal specifically detects Carbonic Anhydrase XIV/CA14 in Human samples. It is validated for Western Blot, ELISA.Specifications
Carbonic Anhydrase XIV/CA14 | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1 | |
Recombinant Human Carbonic Anhydrase XIV/CA14 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
4 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
0.2 um filtered solution in PBS | |
23632 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title