Learn More
ACROBiosystems Recombinant Protein;Human Carbonic Anhydrase II / CA2 Protein;HEK293;100UG;CA2,CA2,CA-II,CAII,Car2,CA-2,Carbonic Anhydrase II
Supplier:  ACROBiosystems CA2H5228100UG
Encompass_Preferred
Recombinant Protein;Human Carbonic Anhydrase II / CA2 Protein;100UG;Human CA2, His Tag (CA2-H5228) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Lys 260 (Accession # NP_000058.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30 kDa. The protein migrates as 30 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8990
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.