Alfa Aesar™ Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I), Rh 21%

Hydroformylation

Manufacturer: Alfa Aesar™ 1045806

Quantity 5g

 View more versions of this product

Catalog No. AA1045806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25470-96-6
    Formula Weight 492.32
    MDL Number MFCD00064611
    Merck Index 14,356
    Chemical Name or Material Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I)

    Specifications

    Formula Weight 492.32
    Physical Form Crystals
    MDL Number MFCD00064611
    Solubility Information Soluble in acetone and chlorinated solvents
    TSCA TSCA
    CAS 25470-96-6
    Merck Index 14,356
    Color Yellow
    Concentration Rh 21%
    Quantity 5g
    Chemical Name or Material Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit