Alfa Aesar™ Carbonylchlorobis(triphenylphosphine)rhodium(I), Rh 14.9% min

Manufacturer: Alfa Aesar™ 3982201

Quantity 0.25g

 View more versions of this product

Catalog No. AA3982201

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13938-94-8
    Linear Formula RhCl(CO)[P(C6H5)3]2
    Formula Weight 690.94
    MDL Number MFCD00044273
    Beilstein 4618896
    Synonym Carbonylbis(triphenylphosphine)rhodium(I) chloride
    Chemical Name or Material Carbonylchlorobis(triphenylphosphine)rhodium(I)

    Specifications

    Formula Weight 690.94
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Odor Odorless
    Linear Formula RhCl(CO)[P(C6H5)3]2
    CAS 13938-94-8
    Synonym Carbonylbis(triphenylphosphine)rhodium(I) chloride
    Color Yellow
    Concentration Rh 14.9% min.
    Physical Form Crystalline
    MDL Number MFCD00044273
    Solubility Information Soluble in acetone, chloroform, ethanol
    Quantity 0.25g
    Beilstein 4618896
    Melting Point 224° to 227°C (dec.)
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 1 Warning
    Chemical Name or Material Carbonylchlorobis(triphenylphosphine)rhodium(I)

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit