Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonyldihydridotris(triphenylphosphine)ruthenium(II), White crystalline, 99%, Alfa Aesar™
Click to view available options
Quantity:
0.5g
2g
10g
Specifications
Color | White |
Melting Point | 161°C to 163°C |
Sensitivity | Light and moisture sensitive |
Quantity | 2g |
Assay Percent Range | 99% |
Solubility Information | Insoluble in water. |
Formula Weight | 917.97 |
Physical Form | Crystals |
Chemical Name or Material | Carbonyldihydridotris(triphenylphosphine)ruthenium(II) |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen