Alfa Aesar™ Carbonylhydridotris(triphenylphosphine)iridium(I), Ir 18.6% min

Manufacturer: Alfa Aesar™ 4100506

Quantity 5g

 View more versions of this product

Catalog No. AA4100506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17250-25-8
    Linear Formula IrH(CO)[P(C6H5)3]3
    Formula Weight 1008.11
    MDL Number MFCD00015525
    Synonym Hydridocarbonyltris(triphenylphosphine)iridium(I); Carbonyltris(triphenylphosphine)iridium(I) hydride
    Chemical Name or Material Carbonylhydridotris(triphenylphosphine)iridium(I)

    Specifications

    Formula Weight 1008.11
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula IrH(CO)[P(C6H5)3]3
    Solubility Information Soluble in chloroform and toluene
    Sensitivity Moisture Sensitive
    Quantity 5g
    Physical Form Powder
    MDL Number MFCD00015525
    Health Hazard 1 Danger
    Melting Point 170°C
    Synonym Hydridocarbonyltris(triphenylphosphine)iridium(I); Carbonyltris(triphenylphosphine)iridium(I) hydride
    Color Pale yellow
    TSCA TSCA
    CAS 17250-25-8
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Concentration Ir 18.6% min.
    Chemical Name or Material Carbonylhydridotris(triphenylphosphine)iridium(I)

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit