Alfa Aesar™ Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min

Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation

Manufacturer: Alfa Aesar™ 1001677

Quantity 0.1g

 View more versions of this product

Catalog No. AA1001677

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17185-29-4
    Linear Formula RhH(CO)[P(C6H5)3]3
    Formula Weight 918.8
    MDL Number MFCD00151644
    Merck Index 14,9758
    Synonym Tris(triphenylphosphine)rhodium carbonyl hydride; Carbonyltris(triphenylphosphine)rhodium(I) hydride
    Chemical Name or Material Carbonylhydridotris(triphenylphosphine)rhodium(I)

    Specifications

    Formula Weight 918.8
    Melting Point 138°C (dec.)
    Synonym Tris(triphenylphosphine)rhodium carbonyl hydride; Carbonyltris(triphenylphosphine)rhodium(I) hydride
    Concentration Rh 10.0% min.
    Quantity 0.1g
    TSCA TSCA
    MDL Number MFCD00151644
    CAS 17185-29-4
    Merck Index 14,9758
    Density 1.33g/mL
    Odor Odorless
    Physical Form Powder
    Solubility Information Soluble in hydrocarbons (e.g. benzene and toluene) with dissociation of phosphine ligands
    Color Yellow
    Sensitivity Moisture Sensitive
    Linear Formula RhH(CO)[P(C6H5)3]3
    Chemical Name or Material Carbonylhydridotris(triphenylphosphine)rhodium(I)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit