Learn More
Carbonyltris(triphenylphosphine)rhodium(I)hydride, 97%, ACROS Organics™
Specifications
Color | Orange |
Infrared Spectrum | Authentic |
Quantity | 500mg |
Assay Percent Range | 97% |
Linear Formula | {(C6H5)3P}3Rh(CO)H |
Merck Index | 15, 9935 |
Formula Weight | 918.8 |
Physical Form | Powder |
Chemical Name or Material | Carbonyltris(triphenylphosphine)rhodium(I)hydride, 97% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning