Learn More
Thermo Scientific CARBOPAC PA20-FAST-4MM GUARD  DFS Item
Manufacturer:  Thermo Scientific 302748
For Use With:Dionex CarboPac PA20-Fast-4µm Standard Bore Analytical ColumnColumn Chemistry:HydroxideDiameter (Metric): 4mmFlow Rate: 1.0mL/minMax. Pressure:5000 psiMobile Phase Compatibility:0-50% acetonitrile, methanol, or acetoneParticle Size: 6µmpH:0-14Length (Metric):30mmTechnique:Carbohydrate AnalysisAnion Exchange Capacity: 18 µeqColumn Construction:PEEKResin Composition:6µm diameter substrate (ethylvinylbenzene 55% crosslinked with divinylbenzene) agglomerated with 130nm MicroBead difunctional quaternary ammonium ion (5.2% crosslinked)Temperature Range:4-60°CType:Standard Bore Guard
Catalog No. NC1483211