Learn More
MilliporeSigma™ Supelco™ Carbosieve™ SIII Thermal Desorption Tubes
Suitable for use with Air Monitoring
$935.00 - $970.00
Specifications
Product Line Carbosieve™
Material Stainless Steel
Methods EPA TO-2
Matrix Carbosieve™ S-III Carbon Molecular Sieve
Diameter (English) Outer 1/4 in.
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
11-100-6749
View Documents
MilliporeSigma™ Supelco™
20012U
Pre-conditioned
Each for $935.00
Add to cart
11-100-6687
View Documents
MilliporeSigma™ Supelco™
29743U
Pre-conditioned/Sealed with Brass Endcaps
Each for $970.00
Add to cart
Specifications
Specifications
Carbosieve™
EPA TO-2
1/4 in.
Air Monitoring
Stainless Steel
Carbosieve™ S-III Carbon Molecular Sieve
3.5 in.
Desorption Tube
SDS
Carbosieve is a registered trademark of Sigma-Aldrich Co. LLC