Learn More
Promega Carboxy Flexi™ Cloning System and Vectors
These vectors are used for inducible expression of HQ--tagged fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter.
Manufacturer:  Promega C8521
 View more versions of this product
Catalog No. PRC8521
Add to cart
Description
Description
  • Protein Expression Vectors with HQ Tag for Ni-Affinity Purification
  • Express N- or C-terminal HQ-tagged proteins in T7 promoter expression systems
  • Choose from a variety of expression systems and fusion tag orientations and then transfer as required
  • Barnase insert decreases the number of background colonies for efficient transfer of ORFs
Order Info
Kit supplied with all components necesssary for 50 transfer reactions; individual vectors sold separately
Specifications
Specifications
Transferring Protein Coding Regions From Native Or N Terminal Flexi Vectors To C Terminal Flexi Vectors
pFN6K (HQ) Flexi Vector
pFN6K (HQ) Flexi Vector
−30 to −10°C
Carboxy Flexi™ Cloning System
Fluid
20 μg