Learn More
Tocris Bioscience™ Carboxy-PTIO, potassium salt Available on GSA/VA Contract for Federal Government customers only.
Stable, water-soluble deactivator of NO
Manufacturer:  Tocris Bioscience™ 0772/10
 View more versions of this product
Catalog No. 07-721-0
Add to cart
Description
Description
A stable, water-soluble free radical that reacts stoichiometrically with NO. Almost twice as strong an inhibitor as N(w)-nitroarginine or N-methylarginine.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
2-(4-Carboxyphenyl)-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide, potassium salt
C14H16KN2O4
10mg
148819-94-7
Observed MW: 315.38
Soluble to 25 mM in ethanol and to 10 mM in water
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at -20°C