Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxyl Ester Lipase/CEL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | Carboxyl Ester Lipase/CEL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157922
|
Novus Biologicals
NBP157922 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
Carboxyl Ester Lipase/CEL Polyclonal specifically detects Carboxyl Ester Lipase/CEL in Human samples. It is validated for Western Blot.Specifications
Carboxyl Ester Lipase/CEL | |
Polyclonal | |
Rabbit | |
BAL, bile salt-activated lipase, Bile salt-stimulated lipase, BSDL, BSSLbile salt-dependent lipase, oncofetal isoform, bucelipase, carboxyl ester hydrolase, Carboxyl ester lipase, carboxyl ester lipase (bile salt-stimulated lipase), CEase, CELL, Cholesterol esterase, EC 3.1.1, EC 3.1.1.13, EC 3.1.1.3, FAP, FAPP, fetoacinar pancreatic protein, LIPA, lysophospholipase, pancreatic, MODY8bile-salt-activated lipase, Pancreatic lysophospholipase, Sterol esterase | |
CEL | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
1056 | |
IgG |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title