Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxymethylcellulose Sodium, High Viscosity, USP, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9004-32-4
C8H16NaO8
263.20
DPXJVFZANSGRMM-UHFFFAOYNA-N
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
9004-32-4
C8H16NaO8
263.20
DPXJVFZANSGRMM-UHFFFAOYNA-N
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

CA194, 9004-32-4

Spectrum™ Carboxymethylcellulose Sodium, High Viscosity, Viscosity of 1 Percent Aqueous Solution @ 25 DEG C: 1500-3000 cP, USP is often used as a thickener and stabilizer. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Loss on Drying 10%
Packaging Fiber Drum
Quantity 45 kg