Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase A1/CPA1 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI2A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270431AF700

Catalog No. N270431A700


Add to cart

Description

Description

Carboxypeptidase A1/CPA1 Monoclonal antibody specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Carboxypeptidase A1/CPA1
OTI2A3
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
CPA1
Recombinant protein expressed in E.coli corresponding to amino acids 1-419 of human Carboxypeptidase A1/CPA1.
Immunogen affinity purified
Mast Cell Markers
Primary
1357.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit